MAT379356469 Matrix Scientific

(2E)-2-[(Dimethylamino)methylene]-cycloheptanone hydrochloride

  • MDL number MFCD22421989
  • Cas Number : 1417368-42-3
  • Molecular Weight : 203.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080612.html

Safety Information

MSDS http://www.matrixscientific.com/080612.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart