MAT379356659 Matrix Scientific

N-[Amino(imino)methyl]-4-methylpiperazine-1-carboximidamide

  • MDL number MFCD15204029
  • Cas Number : 18413-26-8
  • Molecular Weight : 184.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080802.html

Safety Information

MSDS http://www.matrixscientific.com/080802.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart