MAT379356668 Matrix Scientific

3,4-Dihydroisoquinoline-2(1H)-carboximidamide sulfate

  • MDL number MFCD00941496
  • Cas Number : 2514-89-8
  • Molecular Weight : 273.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080811.html

Safety Information

MSDS http://www.matrixscientific.com/080811.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart