MAT379356698 Matrix Scientific

N-[4-(4-Chlorophenyl)-1,3-thiazol-2-yl]guanidine

  • MDL number MFCD02102731
  • Cas Number : 7120-02-7
  • Molecular Weight : 252.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/080841.html

Safety Information

MSDS http://www.matrixscientific.com/080841.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart