MAT379357038 Matrix Scientific

2-Chloroaniline hydrochloride

  • MDL number MFCD00053282
  • Cas Number : 137-04-2
  • Molecular Weight : 164.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220�bl.)
Purity 97
Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/081172.html

Safety Information

MSDS http://www.matrixscientific.com/081172.html
pictograms Toxic/Stored under Argon/Hygroscopic
Availability details for
Quantity
  • Availability
  • Add to Cart