MAT379357139 Matrix Scientific

2-(4-Fluorophenoxy)pyridin-3-amine

  • MDL number MFCD00067803
  • Cas Number : 175135-64-5
  • Molecular Weight : 204.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/081274.html

Safety Information

MSDS http://www.matrixscientific.com/081274.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart