MAT379357236 Matrix Scientific

5-Aminopyrimidine-2,4,6(1H,3H,5H)-trione(Uramil)

  • MDL number MFCD00023873
  • Cas Number : 118-78-5
  • Molecular Weight : 143.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/081371.html

Safety Information

MSDS http://www.matrixscientific.com/081371.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart