MAT379357615 Matrix Scientific

9H-Pyrido[3,4-b]indole-1-carboxaldehyde

  • MDL number MFCD14706724
  • Cas Number : 20127-63-3
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/081752.html

Safety Information

MSDS http://www.matrixscientific.com/081752.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart