MAT379358024 Matrix Scientific

3-Pyrazin-2-ylbenzoic acid

  • MDL number MFCD08669903
  • Cas Number : 856905-13-0
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082159.html

Safety Information

MSDS http://www.matrixscientific.com/082159.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart