MAT379358027 Matrix Scientific

4-(5-Chlorothien-2-yl)benzoic acid

  • MDL number MFCD12547358
  • Cas Number : 290326-23-7
  • Molecular Weight : 238.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082162.html

Safety Information

MSDS http://www.matrixscientific.com/082162.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart