MAT379358129 Matrix Scientific

(2E)-3-(2,5-Dihydroxyphenyl)acrylic acid

  • MDL number MFCD00075834
  • Cas Number : 636-01-1
  • Molecular Weight : 180.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082266.html

Safety Information

MSDS http://www.matrixscientific.com/082266.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart