MAT379358143 Matrix Scientific

3-Hydroxy-2,4,6-triiodobenzoic acid

  • MDL number MFCD00020305
  • Cas Number : 53279-72-4
  • Molecular Weight : 515.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/082281.html

Safety Information

MSDS http://www.matrixscientific.com/082281.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart