MAT379358238 Matrix Scientific

Difluoro[3-(trifluoromethyl)-pyridin-2-yl]acetic acid

  • MDL number MFCD12963754
  • Cas Number : 1215579-44-4
  • Molecular Weight : 241.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082376.html

Safety Information

MSDS http://www.matrixscientific.com/082376.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart