MAT379358347 Matrix Scientific

2-(Cyclobutylamino)isonicotinic acid

  • MDL number MFCD16747175
  • Cas Number : 1251225-65-6
  • Molecular Weight : 192.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082485.html

Safety Information

MSDS http://www.matrixscientific.com/082485.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart