MAT379358427 Matrix Scientific

2-(4-Methoxybenzyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD02102515
  • Cas Number : 477872-94-9
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082565.html

Safety Information

MSDS http://www.matrixscientific.com/082565.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart