MAT379358533 Matrix Scientific

3-Benzyl-1,2,4-oxadiazole-5-carboxylic acid

  • MDL number MFCD07379382
  • Cas Number : 1159694-86-6
  • Molecular Weight : 204.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082671.html

Safety Information

MSDS http://www.matrixscientific.com/082671.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart