MAT379358578 Matrix Scientific

1,2,3,4-Tetrahydroisoquinoline-4-carboxylic acid

  • MDL number MFCD08236715
  • Cas Number : 116140-19-3
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-238
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/082716.html

Safety Information

MSDS http://www.matrixscientific.com/082716.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart