MAT379358823 Matrix Scientific

6,7-Dihydro-5H-pyrrolo[1,2-a]-imidazole-3-carboxylic acid

  • MDL number MFCD08235234
  • Cas Number : 914637-68-6
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/082961.html

Safety Information

MSDS http://www.matrixscientific.com/082961.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart