MAT379358827 Matrix Scientific

7,7-Dimethyl-5-oxo-5,6,7,8-tetrahydroquinoline-3-carboxylic acid

  • MDL number MFCD09414723
  • Cas Number : 1086375-47-4
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/082965.html

Safety Information

MSDS http://www.matrixscientific.com/082965.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart