MAT379358877 Matrix Scientific

4-(Acetylamino)butanoic acid

  • MDL number MFCD00020552
  • Cas Number : 3025-96-5
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-131
Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/083015.html

Safety Information

MSDS http://www.matrixscientific.com/083015.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart