MAT379358947 Matrix Scientific

1-Phenylcyclobutanecarbonyl chloride

  • MDL number MFCD09903688
  • Cas Number : 4620-67-1
  • Molecular Weight : 194.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 15-18
Purity 95
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 84-85�8 Torr
Product Link http://www.matrixscientific.com/083082.html

Safety Information

MSDS http://www.matrixscientific.com/083082.html
pictograms Corrosive/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart