MAT379359197 Matrix Scientific

Methyl 4-amino-3-iodo-5-methylbenzoate

  • MDL number MFCD10703532
  • Cas Number : 180624-11-7
  • Molecular Weight : 291.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083333.html

Safety Information

MSDS http://www.matrixscientific.com/083333.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart