MAT379359226 Matrix Scientific

Methyl 3-phenoxybenzoate

  • MDL number MFCD00017192
  • Cas Number : 50789-43-0
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083362.html

Safety Information

MSDS http://www.matrixscientific.com/083362.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart