MAT379359238 Matrix Scientific

Ethyl isonicotinate 1-oxide

  • MDL number MFCD06204693
  • Cas Number : 14906-37-7
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083374.html

Safety Information

MSDS http://www.matrixscientific.com/083374.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart