MAT379359306 Matrix Scientific

Methyl 3-amino-4-(isopropylsulfonyl)-thiophene-2-carboxylate

  • MDL number MFCD00067930
  • Cas Number : 175201-72-6
  • Molecular Weight : 263.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083442.html

Safety Information

MSDS http://www.matrixscientific.com/083442.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart