MAT379359345 Matrix Scientific

Methyl 5-(hydroxymethyl)isoxazole-3-carboxylate

  • MDL number MFCD14706527
  • Cas Number : 139297-55-5
  • Molecular Weight : 157.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083481.html

Safety Information

MSDS http://www.matrixscientific.com/083481.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart