MAT379359546 Matrix Scientific

Isopropyl 2-bromo-2-methylpropanoate

  • MDL number MFCD00055235
  • Cas Number : 51368-55-9
  • Molecular Weight : 209.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083682.html

Safety Information

MSDS http://www.matrixscientific.com/083682.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart