MAT379359663 Matrix Scientific

2-(4-Bromophenoxy)-6-(trifluoromethyl)pyridine

  • MDL number MFCD10703540
  • Cas Number : 1086376-15-9
  • Molecular Weight : 318.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/083800.html

Safety Information

MSDS http://www.matrixscientific.com/083800.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart