MAT379360059 Matrix Scientific

3-Chloro-4-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD04972758
  • Cas Number : 948014-51-5
  • Molecular Weight : 210.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-58
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/084191.html

Safety Information

MSDS http://www.matrixscientific.com/084191.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart