MAT379360460 Matrix Scientific

2-Propylbenzene-1,3-diol

  • MDL number MFCD00100568
  • Cas Number : 13331-19-6
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-105
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/084592.html

Safety Information

MSDS http://www.matrixscientific.com/084592.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart