MAT379360824 Matrix Scientific

4,6-Di(tert-butyl)benzene-1,3-diol

  • MDL number MFCD00044513
  • Cas Number : 5374-06-1
  • Molecular Weight : 222.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-123
Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/084956.html

Safety Information

MSDS http://www.matrixscientific.com/084956.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart