MAT379360957 Matrix Scientific

1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone

  • MDL number MFCD12404912
  • Cas Number : 1216474-77-9
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/085088.html

Safety Information

MSDS http://www.matrixscientific.com/085088.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart