MAT379360958 Matrix Scientific

1-[6-(3-Chlorophenyl)pyridin-3-yl]ethanone

  • MDL number MFCD12404913
  • Cas Number : 1216636-84-8
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/085089.html

Safety Information

MSDS http://www.matrixscientific.com/085089.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart