MAT379360959 Matrix Scientific

1-[6-(4-Methoxyphenyl)pyridin-3-yl]ethanone

  • MDL number MFCD12404914
  • Cas Number : 1215576-19-4
  • Molecular Weight : 227.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/085090.html

Safety Information

MSDS http://www.matrixscientific.com/085090.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart