MAT379361137 Matrix Scientific

3-Methylthiophene-2-carbonitrile

  • MDL number MFCD00052086
  • Cas Number : 55406-13-8
  • Molecular Weight : 123.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 91-93�mm
Product Link http://www.matrixscientific.com/085269.html

Safety Information

MSDS http://www.matrixscientific.com/085269.html
pictograms Toxic
Availability details for
Quantity
  • Availability
  • Add to Cart