MAT379361516 Matrix Scientific

1-Benzyl-4-hydroxypiperidine-4-carbonitrile

  • MDL number MFCD00023752
  • Cas Number : 6094-60-6
  • Molecular Weight : 216.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-102
Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/085649.html

Safety Information

MSDS http://www.matrixscientific.com/085649.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart