MAT379361551 Matrix Scientific

1-(5-Chloro-2-methoxyphenyl)-piperazine hydrochloride

  • MDL number MFCD00674132
  • Cas Number : 99857-72-4
  • Molecular Weight : 263.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/085684.html

Safety Information

MSDS http://www.matrixscientific.com/085684.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart