MAT379361775 Matrix Scientific

2-Methoxy-1-naphthonitrile

  • MDL number MFCD00003867
  • Cas Number : 16000-39-8
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/085908.html

Safety Information

MSDS http://www.matrixscientific.com/085908.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart