MAT379361928 Matrix Scientific

Cyanomethyl ethanethioate

  • MDL number MFCD00173920
  • Cas Number : 59463-56-8
  • Molecular Weight : 115.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086064.html

Safety Information

MSDS http://www.matrixscientific.com/086064.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart