MAT379362050 Matrix Scientific

3-(3-Cyanophenyl)-1-propene

  • MDL number MFCD01319565
  • Cas Number : 61463-62-5
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086202.html

Safety Information

MSDS http://www.matrixscientific.com/086202.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart