MAT379362129 Matrix Scientific

3-(5-Bromo-2-thienyl)-1-propene

  • MDL number MFCD09801334
  • Cas Number : 159013-60-2
  • Molecular Weight : 203.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/086282.html

Safety Information

MSDS http://www.matrixscientific.com/086282.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart