MAT379362150 Matrix Scientific

5-(4-Chlorophenyl)-2-methylpyridine

  • MDL number MFCD13152444
  • Cas Number : 23148-35-8
  • Molecular Weight : 203.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086303.html

Safety Information

MSDS http://www.matrixscientific.com/086303.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart