MAT379362151 Matrix Scientific

4-(4-Chlorophenyl)-2-methylpyridine

  • MDL number MFCD13152445
  • Cas Number : 23148-58-5
  • Molecular Weight : 203.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-70
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086304.html

Safety Information

MSDS http://www.matrixscientific.com/086304.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart