MAT379362154 Matrix Scientific

3-(4-Bromo-2-fluorophenyl)pyridine

  • MDL number MFCD13152460
  • Cas Number : 1187163-18-3
  • Molecular Weight : 252.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-52
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086307.html

Safety Information

MSDS http://www.matrixscientific.com/086307.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart