MAT379362157 Matrix Scientific

4-(3-Thienyl)isoquinoline

  • MDL number MFCD02260315
  • Cas Number : 112370-12-4
  • Molecular Weight : 211.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-200
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086310.html

Safety Information

MSDS http://www.matrixscientific.com/086310.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart