MAT379362158 Matrix Scientific

Ethyl 5-(4-methyl-3-pyridyl)-2-furoate

  • MDL number MFCD13152496
  • Cas Number : 1187163-61-6
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-40
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086311.html

Safety Information

MSDS http://www.matrixscientific.com/086311.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart