MAT379362159 Matrix Scientific

5-(4-Methyl-pyridin-3-yl)-thiophene-2-carboxylic acid ethyl ester

  • MDL number MFCD13152509
  • Cas Number : 1187163-59-2
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086312.html

Safety Information

MSDS http://www.matrixscientific.com/086312.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart