MAT379362165 Matrix Scientific

5-Bromo-[2,3']bipyridinyl

  • MDL number MFCD09037420
  • Cas Number : 774-53-8
  • Molecular Weight : 235.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-77
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086319.html

Safety Information

MSDS http://www.matrixscientific.com/086319.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart