MAT379362166 Matrix Scientific

5-Pyridin-2-yl-pyrimidine

  • MDL number MFCD13152524
  • Cas Number : 64858-29-3
  • Molecular Weight : 157.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-133
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086320.html

Safety Information

MSDS http://www.matrixscientific.com/086320.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart