MAT379362167 Matrix Scientific

6'-Chloro-5-methyl-[2,3']bipyridinyl

  • MDL number MFCD13152543
  • Cas Number : 1187168-72-4
  • Molecular Weight : 204.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/086321.html

Safety Information

MSDS http://www.matrixscientific.com/086321.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart